6-Fluorochromone

Supplier: Apollo Scientific

Synonyms: 6-Fluoro-4H-chromen-4-one

PC0953-1G PC0953-5G
APOSPC0953-1GEA 80 GBP
APOSPC0953-1G APOSPC0953-5G
6-Fluorochromone
6-Fluorochromone
Formula: C₉H₅FO₂
MW: 164,14 g/mol
Melting Pt: 166…169 °C
Storage Temperature: Ambient
MDL Number: MFCD03094002
CAS Number: 105300-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR