3-Fluoro-2-methylbenzaldehyde 96%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Fluoro-o-tolualdehyde, 3-Fluoro-2-tolualdehyde, 3-Fluoro-o-tolylaldehyde
PC0881-100G
PC0881-25G
PC0881-5G
APOSPC0881-100GEA
171
GBP
APOSPC0881-100G
APOSPC0881-25G
APOSPC0881-5G
3-Fluoro-2-methylbenzaldehyde 96%
3-Fluoro-2-methylbenzaldehyde
Formula:
C₈H₇FO Boiling Pt: 92…93 °C/20mmHg Density: 1,16 g/cm³ (25 °C) Flash Pt: 81 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00075256 CAS Number: 147624-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...