(S)-(+)-ɑ-(Trifluoromethyl)benzyl alcohol 99%

Supplier: Apollo Scientific
Warning

Synonyms: (S)-2,2,2-Trifluoro-1-phenylethanol, (S)-(+)-1-Phenyl-2,2,2-trifluoroethanol

PC0769-1G PC0769-500MG PC0769-250MG
APOSPC0769-1GEA 57.7 GBP
APOSPC0769-1G APOSPC0769-500MG APOSPC0769-250MG
(S)-(+)-ɑ-(Trifluoromethyl)benzyl alcohol 99%
(S)-(+)-alpha-(Trifluoromethyl)benzyl alcohol
Formula: C₈H₇F₃O
MW: 176,14 g/mol
Boiling Pt: 73…76 °C
Density: 1,3 g/cm³ (25 °C)
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00077845
CAS Number: 340-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR