1,2,3,3,3-Pentafluoropropene 97%

Supplier: Apollo Scientific
Warning

PC0752-25G PC0752-5G
APOSPC0752-25GEA 709 GBP
APOSPC0752-25G APOSPC0752-5G
1,2,3,3,3-Pentafluoropropene 97%
1,2,3,3,3-Pentafluoropropene
Formula: C₃HF₅
MW: 132,03 g/mol
Boiling Pt: –18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02262202
CAS Number: 2252-83-7
UN: 3163
ADR: 2.2,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR