4-Fluoro-N-(4-fluorobenzylidene)aniline

Supplier: Apollo Scientific

PC0720-1G PC0720-5G
APOSPC0720-1GEA 72.2 GBP
APOSPC0720-1G APOSPC0720-5G
4-Fluoro-N-(4-fluorobenzylidene)aniline
4-Fluoro-N-(4-fluorobenzylidene)aniline
Formula: C₁₃H₉F₂N
MW: 217,22 g/mol
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00017950
CAS Number: 39769-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR