Carmofur

Supplier: Apollo Scientific

Synonyms: 5-Fluoro-N-hexyl-2,4-dioxo-3,4-dihydropyrimidine-1(2H)-carboxamide, 5-Fluoro-1-hexylcarbamoyluracil

PC0659-1G PC0659-5G
APOSPC0659-1GEA 72.2 GBP
APOSPC0659-1G APOSPC0659-5G
Carmofur
Carmofur
Formula: C₁₁H₁₆FN₃O₃
MW: 257,27 g/mol
Melting Pt: 110…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00866284
CAS Number: 61422-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR