1,1,1,2,3-Pentafluoropropane 99%
Supplier: Apollo Scientific
|
Warning
|
PC0638-25G
PC0638-5G
APOSPC0638-25GEA
1120
GBP
APOSPC0638-25G
APOSPC0638-5G
1,1,1,2,3-Pentafluoropropane 99%
1,1,1,2,3-Pentafluoropropane
Formula:
C₃H₃F₅ Boiling Pt: 23 °C Density: 1,267 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00236624 CAS Number: 431-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...