(S)-(-)-4-Fluoro-α-methylbenzyl alcohol 98%

Supplier: Apollo Scientific
Warning

Synonyms: (S)-1-(4-Fluorophenyl)ethanol, (1S)-1-(4-Fluorophenyl)ethan-1-ol

PC0610-1G PC0610-250MG
APOSPC0610-1GEA 191 GBP
APOSPC0610-1G APOSPC0610-250MG
(S)-(-)-4-Fluoro-α-methylbenzyl alcohol 98%
(S)-(-)-4-Fluoro-α-methylbenzyl alcohol
Formula: C₈H₉FO
MW: 140,16 g/mol
Density: 1,111 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03092998
CAS Number: 101219-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR