N-Fmoc-2-fluoro-L-phenylalanine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-fluorophenyl)propanoic acid, Fmoc-Phe(2-F)-OH
PC0426-10G
PC0426-25G
APOSPC0426-10GEA
120
GBP
APOSPC0426-10G
APOSPC0426-25G
N-Fmoc-2-fluoro-L-phenylalanine
N-Fmoc-2-fluoro-L-phenylalanine
Formula:
C₂₄H₂₀FNO₄ MW: 405,425 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00672552 CAS Number: 205526-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...