4-Fluoro-3-methylbenzyl bromide

Supplier: Apollo Scientific
Danger

PC0363-10G PC0363-1G PC0363-5G PC0363-25G
APOSPC0363-10GEA 255 GBP
APOSPC0363-10G APOSPC0363-1G APOSPC0363-5G APOSPC0363-25G
4-Fluoro-3-methylbenzyl bromide
4-Fluoro-3-methylbenzyl bromide
Formula: C₈H₈BrF
Boiling Pt: 80 °C/3mmHg
Storage Temperature: Ambient
MDL Number: MFCD01631425
CAS Number: 261951-70-6
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR