4-Fluoro-3-methylbenzylamine 97%
Supplier: Apollo Scientific
|
Danger
|
PC0357-100G
PC0357-25G
PC0357-5G
APOSPC0357-100GEA
157
GBP
APOSPC0357-100G
APOSPC0357-25G
APOSPC0357-5G
4-Fluoro-3-methylbenzylamine 97%
4-Fluoro-3-methylbenzylamine
Formula:
C₈H₁₀FN Boiling Pt: 145 °C Density: 1,059 g/cm³ (25 °C) Flash Pt: 68 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631490 CAS Number: 261951-68-2 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...