4-Fluoro-3-methylbenzaldehyde
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Fluoro-m-tolylaldehyde, 4-Fluoro-3-tolualdehyde, 4-Fluoro-m-tolualdehyde
PC0340-100G
PC0340-25G
PC0340-5G
APOSPC0340-100GEA
169
GBP
APOSPC0340-100G
APOSPC0340-25G
APOSPC0340-5G
4-Fluoro-3-methylbenzaldehyde
4-Fluoro-3-methylbenzaldehyde
Formula:
C₈H₇FO MW: 138,14 g/mol Boiling Pt: 208 °C Density: 1,132 g/cm³ (25 °C) Flash Pt: 79 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631431 CAS Number: 135427-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...