4-[5-(4-Fluorophenyl)-1,3,4-oxadiazol-2-yl]piperidine 95+%
Supplier: Apollo Scientific
|
Warning
|
PC0207-250MG
PC0207-100MG
APOSPC0207-250MGEA
55.1
GBP
APOSPC0207-250MG
APOSPC0207-100MG
4-[5-(4-Fluorophenyl)-1,3,4-oxadiazol-2-yl]piperidine 95+%
4-[5-(4-Fluorophenyl)-1,3,4-oxadiazol-2-yl]piperidine
Formula:
C₁₃H₁₄FN₃O MW: 247,27 g/mol Melting Pt: 94…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD03424488 CAS Number: 493024-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...