2-Chloro-5-(trifluoromethyl)benzyl alcohol

Supplier: Apollo Scientific
Warning

PC0191-100G PC0191-25G PC0191-5G
APOSPC0191-100GEA 168 GBP
APOSPC0191-100G APOSPC0191-25G APOSPC0191-5G
2-Chloro-5-(trifluoromethyl)benzyl alcohol
2-Chloro-5-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆ClF₃O
MW: 210.58 g/mol
Melting Pt: 57…60 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01631469
CAS Number: 64372-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR