2-Chloro-5-(trifluoromethyl)benzamide 97%
Supplier: Apollo Scientific
PC0186-10G
PC0186-5G
PC0186-1G
APOSPC0186-10GEA
145
GBP
APOSPC0186-10G
APOSPC0186-5G
APOSPC0186-1G
2-Chloro-5-(trifluoromethyl)benzamide 97%
2-Chloro-5-(trifluoromethyl)benzamide
Formula:
C₈H₅ClF₃NO Storage Temperature: Ambient |
MDL Number:
MFCD01631467 CAS Number: 20566-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...