3-Chloro-2-fluorophenylacetonitrile
Supplier: Apollo Scientific
|
Danger
|
PC0136-1G
PC0136-5G
PC0136-10G
APOSPC0136-1GEA
15.8
GBP
APOSPC0136-1G
APOSPC0136-5G
APOSPC0136-10G
3-Chloro-2-fluorophenylacetonitrile
3-Chloro-2-fluorophenylacetonitrile
Formula:
C₈H₅ClFN MW: 169,5800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01631552 CAS Number: 261762-98-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...