2-Chloro-6-fluorophenol 99+%

Supplier: Apollo Scientific
Danger

PC0129-100G PC0129-25G PC0129-5G
APOSPC0129-100GEA 64.2 GBP
APOSPC0129-100G APOSPC0129-25G APOSPC0129-5G
2-Chloro-6-fluorophenol 99+%
2-Chloro-6-fluorophenol
Formula: C₆H₄ClFO
MW: 146.55 g/mol
Boiling Pt: 199
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD01631574
CAS Number: 2040-90-6
EINECS: 433-890-8
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR