2,5-Dimethoxybenzaldehyde 98%
Supplier: Apollo Scientific
|
Warning
|
OR9939-25G
OR9939-5G
APOSOR9939-25GEA
39.4
GBP
APOSOR9939-25G
APOSOR9939-5G
2,5-Dimethoxybenzaldehyde 98%
2,5-Dimethoxybenzaldehyde
Formula:
C₉H₁₀O₃ MW: 166,18 g/mol Boiling Pt: 146 °C/10mmHg Melting Pt: 46…51 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003314 CAS Number: 93-02-7 EINECS: 202-211-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...