tert-Butyl-4-(4-aminobenzoyl)tetrahydro-1(2H)-pyrazinecarboxylate

Supplier: Apollo Scientific

Synonyms: tert-butyl 4-(4-aminobenzoyl)piperazine-1-carboxylate

OR9915-1G OR9915-500MG
APOSOR9915-1GEA 408 GBP
APOSOR9915-1G APOSOR9915-500MG
tert-Butyl-4-(4-aminobenzoyl)tetrahydro-1(2H)-pyrazinecarboxylate
tert-Butyl-4-(4-aminobenzoyl)tetrahydro-1(2H)-pyrazinecarboxylate
Formula: C₁₆H₂₃N₃O₃
MW: 305,38 g/mol
Melting Pt: 159…160 °C
Storage Temperature: Ambient
MDL Number: MFCD06659531
CAS Number: 350684-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR