2-Amino-4'-chloroacetophenone hydrochloride 95%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Chlorophenacylamine hydrochloride

OR9907-10G OR9907-25G OR9907-5G
APOSOR9907-10GEA 211 GBP
APOSOR9907-10G APOSOR9907-25G APOSOR9907-5G
2-Amino-4'-chloroacetophenone hydrochloride 95%
2-Amino-4'-chloroacetophenone hydrochloride
Formula: C₈H₉Cl₂NO
Melting Pt: 262 °C
Storage Temperature: Ambient
MDL Number: MFCD00193061
CAS Number: 5467-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR