3-Benzylaniline
Supplier: Apollo Scientific
|
Warning
|
OR9849-5G
OR9849-1G
APOSOR9849-5GEA
748
GBP
APOSOR9849-5G
APOSOR9849-1G
3-Benzylaniline
3-Benzylaniline
Formula:
C₁₃H₁₃N MW: 183,25 g/mol Melting Pt: 48…52 °C Flash Pt: 44 °C Storage Temperature: Ambient |
MDL Number:
MFCD01310782 CAS Number: 61424-26-8 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...