5-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-yl)-2,3-dihydrothieno[3,4-b][1,4]dioxane 97%

Supplier: Apollo Scientific
Warning

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydrothieno[3,4-b][1,4]dioxine, 2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-boronic acid Pinacol ester, 2-(2,3-Dihydrothieno[3,4-b][1,4]dioxin-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,4-Ethylenedioxythiophene-2-boronic acid Pinacol ester

OR9767-100MG OR9767-1G OR9767-250MG
APOSOR9767-100MGEA 106 GBP
APOSOR9767-100MG APOSOR9767-1G APOSOR9767-250MG
5-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-yl)-2,3-dihydrothieno[3,4-b][1,4]dioxane 97%
5-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-yl)-2,3-dihydrothieno[3,4-b][1,4]dioxane
Formula: C₁₂H₁₇BO₄S
MW: 268,14 g/mol
Melting Pt: 92…95 °C
Storage Temperature: Ambient
MDL Number: MFCD08060541
CAS Number: 250726-93-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR