(1S)-1-(4-Pyridyl)ethylamine 95%
Supplier: Apollo Scientific
Synonyms:
(S)-1-(4-Pyridyl)ethylamine
OR965857-1G
OR965857-250MG
APOSOR965857-1GEA
215
GBP
APOSOR965857-1G
APOSOR965857-250MG
(1S)-1-(4-Pyridyl)ethylamine 95%
(1S)-1-(4-Pyridyl)ethylamine
Formula:
C₇H₁₀N₂ MW: 122,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09256819 CAS Number: 27854-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...