4-Amino-3,5-dimethylbenzonitrile 97%

Supplier: Apollo Scientific

OR965815-1G OR965815-250MG OR965815-5G
APOSOR965815-1GEA 49.9 GBP
APOSOR965815-1G APOSOR965815-250MG APOSOR965815-5G
4-Amino-3,5-dimethylbenzonitrile 97%
4-Amino-3,5-dimethylbenzonitrile
Formula: C₉H₁₀N₂
MW: 146,1891 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07369841
CAS Number: 74896-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR