Cyclobutanol
Supplier: Apollo Scientific
OR965150-10G
OR965150-25G
APOSOR965150-10GEA
125
GBP
APOSOR965150-10G
APOSOR965150-25G
Cyclobutanol
Cyclobutanol
Formula:
C₄H₈O MW: 72,11 g/mol Flash Pt: 21 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001318 CAS Number: 2919-23-5 UN: 1987 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...