Ethyl thiazole-2-carboxylate 95%
Supplier: Apollo Scientific
OR964945-1G
OR964945-250MG
APOSOR964945-1GEA
74.8
GBP
APOSOR964945-1G
APOSOR964945-250MG
Ethyl thiazole-2-carboxylate 95%
Ethyl thiazole-2-carboxylate
Formula:
C₆H₇NO₂S MW: 157,1900 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06204689 CAS Number: 14527-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...