H-D-Pro-OtBu 98%

Supplier: Apollo Scientific

Synonyms: H-D-Pro-OtBu (syrup)

OR964379-1G OR964379-250MG
APOSOR964379-1GEA 263 GBP
APOSOR964379-1G APOSOR964379-250MG
H-D-Pro-OtBu 98%
H-D-Pro-OtBu
Formula: C₉H₁₇NO₂
MW: 171,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00083683
CAS Number: 90071-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR