2-Amino-5-chlorobenzoxazole 97%

Supplier: Apollo Scientific

Synonyms: 5-Chlorobenzo[d]oxazol-2-amine, Zoxazolamine, 2-Amino-5-chloro-1,3-benzoxazole

OR963716-10G OR963716-25G
APOSOR963716-10GEA 171 GBP
APOSOR963716-10G APOSOR963716-25G
2-Amino-5-chlorobenzoxazole 97%
2-Amino-5-chlorobenzoxazole
Formula: C₇H₅ClN₂O
MW: 168,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005770
CAS Number: 61-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR