1-(2-Methylthiazol-4-ylmethyl)piperazine dihydrochloride 95%
Supplier: Apollo Scientific
Synonyms:
1-(2-Methyl-thiazol-4-ylmethyl)-piperazine
OR962370-10G
OR962370-1G
OR962370-5G
APOSOR962370-10GEA
761
GBP
APOSOR962370-10G
APOSOR962370-1G
APOSOR962370-5G
1-(2-Methylthiazol-4-ylmethyl)piperazine dihydrochloride 95%
1-(2-Methylthiazol-4-ylmethyl)piperazine dihydrochloride
Formula:
C₉H₁₇Cl₂N₃S MW: 270,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10007891 CAS Number: 880361-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...