Fmoc-Tle-OH 97%

Supplier: Apollo Scientific

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tert-leucine, (S)-2-(Fmoc-amino)-3,3-dimethylbutyric acid, Fmoc-Tle-OH, Fmoc-α-t-butylglycine, (S)-2-[[[(9H-Fluoren-9-yl)methoxy]carbonyl]amino]-3,3-dimethylbutanoic acid, N-Fmoc-L-tert-leucine, Fmoc-tBu-Gly-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3,3-dimethylbutanoic acid

OR960782-1G OR960782-5G
APOSOR960782-1GEA 46 GBP
APOSOR960782-1G APOSOR960782-5G
Fmoc-Tle-OH 97%
Fmoc-Tle-OH
Formula: C₂₁H₂₃NO₄
MW: 353,418 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065649
CAS Number: 132684-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR