3,4-Dihydro-4-oxoquinazoline-6-carboxylic acid 95%

Supplier: Apollo Scientific

OR960752-100MG OR960752-1G OR960752-250MG
APOSOR960752-100MGEA 197 GBP
APOSOR960752-100MG APOSOR960752-1G APOSOR960752-250MG
3,4-Dihydro-4-oxoquinazoline-6-carboxylic acid 95%
3,4-Dihydro-4-oxoquinazoline-6-carboxylic acid
Formula: C₉H₈N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09878505
CAS Number: 33986-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR