1-Benzyl-3,5-dibromo-1H-1,2,4-triazole 95%

Supplier: Apollo Scientific
Warning

OR9607-1G OR9607-5G
APOSOR9607-1GEA 49.9 GBP
APOSOR9607-1G APOSOR9607-5G
1-Benzyl-3,5-dibromo-1H-1,2,4-triazole 95%
1-Benzyl-3,5-dibromo-1H-1,2,4-triazole
Formula: C₉H₇Br₂N₃
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD01568839
CAS Number: 106724-85-0
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR