2-Chloro-3-isothiocyanatopropene 95%
Supplier: Apollo Scientific
OR960670-1G
OR960670-5G
APOSOR960670-1GEA
135
GBP
APOSOR960670-1G
APOSOR960670-5G
2-Chloro-3-isothiocyanatopropene 95%
2-Chloro-3-isothiocyanatopropene
Formula:
C₄H₄ClNS MW: 133.6 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01716868 CAS Number: 14214-31-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...