4-Chloro-2-trimethylsilylpyridine 95%
Supplier: Apollo Scientific
OR960623-1G
OR960623-250MG
APOSOR960623-1GEA
0
GBP
APOSOR960623-1G
APOSOR960623-250MG
4-Chloro-2-trimethylsilylpyridine 95%
4-Chloro-2-trimethylsilylpyridine
Formula:
C₈H₁₂ClNSi MW: 185.73 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09870063 CAS Number: 139585-50-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...