Fmoc-N-Me-Val-OH 97%

Supplier: Apollo Scientific

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-valine, Fmoc-N-Me-Val-OH, N-Fmoc-N-methyl-L-valine

OR960386-1G OR960386-25G OR960386-5G
APOSOR960386-1GEA 87.8 GBP
APOSOR960386-1G APOSOR960386-25G APOSOR960386-5G
Fmoc-N-Me-Val-OH 97%
Fmoc-N-Me-Val-OH
Formula: C₂₁H₂₃NO₄
MW: 353,418 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153395
CAS Number: 84000-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR