4-Methoxypyrimidine 97%

Supplier: Apollo Scientific

OR959609-1G OR959609-5G
APOSOR959609-1GEA 231 GBP
APOSOR959609-1G APOSOR959609-5G
4-Methoxypyrimidine 97%
4-Methoxypyrimidine
Formula: C₅H₆N₂O
MW: 110,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12405815
CAS Number: 6104-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR