Dimethoxymethylphenylsilane 98%

Supplier: Apollo Scientific

Synonyms: Methylphenyldimethoxysilane, Dimethoxy(methyl)(phenyl)silane

OR958491-100G OR958491-25G OR958491-5G
APOSOR958491-100GEA 64.3 GBP
APOSOR958491-100G APOSOR958491-25G APOSOR958491-5G
Dimethoxymethylphenylsilane 98%
Dimethoxymethylphenylsilane
Formula: C₉H₁₄O₂Si
MW: 182,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008343
CAS Number: 3027-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR