Methyl-5-hydroxynicotinate 98%

Supplier: Apollo Scientific
Danger

Synonyms: Methyl 5-hydroxynicotinate, Methyl 5-hydroxy-3-pyridinecarboxylate

OR9567-5G OR9567-25G
APOSOR9567-5GEA 62.9 GBP
APOSOR9567-5G APOSOR9567-25G
Methyl-5-hydroxynicotinate 98%
Methyl 5-hydroxynicotinate
Formula: C₇H₇NO₃
MW: 153,14 g/mol
Melting Pt: 195…197 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191521
CAS Number: 30766-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR