1-Nitrocyclohexene 95%

Supplier: Apollo Scientific

OR956242-1G OR956242-5G OR956242-250MG
APOSOR956242-1GEA 314 GBP
APOSOR956242-1G APOSOR956242-5G APOSOR956242-250MG
1-Nitrocyclohexene 95%
1-Nitrocyclohexene
Formula: C₆H₉NO₂
Storage Temperature: Ambient
CAS Number: 2562-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR