2-(Ethylthio)benzoic acid 95%
Supplier: Apollo Scientific
OR954047-1G
OR954047-250MG
APOSOR954047-1GEA
394
GBP
APOSOR954047-1G
APOSOR954047-250MG
2-(Ethylthio)benzoic acid 95%
2-(Ethylthio)benzoic acid
Formula:
C₉H₁₀O₂S Storage Temperature: Ambient |
MDL Number:
MFCD00094046 CAS Number: 21101-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...