Ethyl-2-chlorothiazole-5-carboxylate 95%

Supplier: Apollo Scientific

Synonyms: Ethyl 2-chlorothiazole-5-carboxylate

OR953268-10G OR953268-1G OR953268-5G
APOSOR953268-10GEA 97.1 GBP
APOSOR953268-10G APOSOR953268-1G APOSOR953268-5G
Ethyl-2-chlorothiazole-5-carboxylate 95%
Ethyl-2-chlorothiazole-5-carboxylate
Formula: C₆H₆ClNO₂S
MW: 191,6353 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08703267
CAS Number: 81449-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR