2,3-Dihydro[1,4]dioxino[2,3-b]pyridin-7-amine 95%

Supplier: Apollo Scientific
Warning

OR952663-100MG OR952663-250MG
APOSOR952663-100MGEA 291 GBP
APOSOR952663-100MG APOSOR952663-250MG
2,3-Dihydro[1,4]dioxino[2,3-b]pyridin-7-amine 95%
2,3-Dihydro[1,4]dioxino[2,3-b]pyridin-7-amine
Formula: C₇H₈N₂O₂
MW: 152,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18374085
CAS Number: 1261365-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR