6-Methoxy-2-naphthol 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Methoxynaphthalen-2-ol, Naproxen Impurity H
OR952419-25G
OR952419-5G
APOSOR952419-25GEA
525
GBP
APOSOR952419-25G
APOSOR952419-5G
6-Methoxy-2-naphthol 95%
6-Methoxy-2-naphthol
Formula:
C₁₁H₁₀O₂ MW: 174,2000 g/mol Melting Pt: 148…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00088677 CAS Number: 5111-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...