9-Boc-3-oxo-9-azabicyclo[3.3.1]nonane 98%

Supplier: Apollo Scientific
Warning

Synonyms: tert-Butyl 3-oxo-9-azabicyclo[3.3.1]nonane-9-carboxylate, 9-Azabicyclo[3.3.1]nonan-3-one, N-BOC protected

OR952038-100G OR952038-25G OR952038-5G
APOSOR952038-100GEA 656 GBP
APOSOR952038-100G APOSOR952038-25G APOSOR952038-5G
9-Boc-3-oxo-9-azabicyclo[3.3.1]nonane 98%
9-Boc-3-oxo-9-azabicyclo[3.3.1]nonane
Formula: C₁₃H₂₁NO₃
MW: 239,32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09997838
CAS Number: 512822-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR