1-Chloro-6-methylisoquinoline 97%
Supplier: Apollo Scientific
OR951758-100MG
OR951758-1G
OR951758-250MG
APOSOR951758-100MGEA
51.1
GBP
APOSOR951758-100MG
APOSOR951758-1G
APOSOR951758-250MG
1-Chloro-6-methylisoquinoline 97%
1-Chloro-6-methylisoquinoline
Formula:
C₁₀H₈ClN MW: 177,6302 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11110400 CAS Number: 209286-73-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...