2-(2-Aminoethoxy)benzonitrile 95%

Supplier: Apollo Scientific

OR951747-1G OR951747-250MG
APOSOR951747-1GEA 328 GBP
APOSOR951747-1G APOSOR951747-250MG
2-(2-Aminoethoxy)benzonitrile 95%
2-(2-Aminoethoxy)benzonitrile
Formula: C₉H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD16495889
CAS Number: 57276-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR