4-Methyltetrahydro-2H-pyran-4-carboxylic acid 95%

Supplier: Apollo Scientific

OR951546-1G OR951546-250MG
APOSOR951546-1GEA 84 GBP
APOSOR951546-1G APOSOR951546-250MG
4-Methyltetrahydro-2H-pyran-4-carboxylic acid 95%
4-Methyltetrahydro-2H-pyran-4-carboxylic acid
Formula: C₇H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD11977785
CAS Number: 233276-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR