Ethyl 1-(aminomethyl)cyclopropanecarboxylate hydrochloride 95%

Supplier: Apollo Scientific

Synonyms: Ethyl-1-(aminomethyl)cyclopropanecarboxylate hydrochloride, 1-(Aminomethyl)-cyclopropanecarboxylic acid ethyl ester hydrochloride

OR951508-1G OR951508-250MG
APOSOR951508-1GEA 120 GBP
APOSOR951508-1G APOSOR951508-250MG
Ethyl 1-(aminomethyl)cyclopropanecarboxylate hydrochloride 95%
Ethyl 1-(aminomethyl)cyclopropanecarboxylate hydrochloride
Formula: C₇H₁₄ClNO₂
MW: 179,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18071250
CAS Number: 362703-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR