Ethyl pyrimidine-2-carboxylate 98%

Supplier: Apollo Scientific

OR951507-10G OR951507-1G OR951507-5G
APOSOR951507-10GEA 81.4 GBP
APOSOR951507-10G APOSOR951507-1G APOSOR951507-5G
Ethyl pyrimidine-2-carboxylate 98%
Ethyl pyrimidine-2-carboxylate
Formula: C₇H₈N₂O₂
MW: 152,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12197317
CAS Number: 42839-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR