3-(2-Aminoethoxy)benzonitrile 95%

Supplier: Apollo Scientific

OR951396-1G OR951396-250MG
APOSOR951396-1GEA 153 GBP
APOSOR951396-1G APOSOR951396-250MG
3-(2-Aminoethoxy)benzonitrile 95%
3-(2-Aminoethoxy)benzonitrile
Formula: C₉H₁₀N₂O
MW: 162,1900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16495888
CAS Number: 120351-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR